ENA372573351 Enamine

3-fluoro-N-phenylaniline

  • MDL number MFCD03094201
  • Cas Number : 500-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112823
Availability details for
Quantity
  • Availability
  • Add to Cart