ENA372573526 Enamine

1-[5-(2,2,2-trifluoroethyl)-1,2,4-oxadiazol-3-yl]methanamine hydrochloride

  • MDL number MFCD22392236
  • Cas Number : 1421601-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112938
Availability details for
Quantity
  • Availability
  • Add to Cart