ENA372573528 Enamine

5-({3-methyl-[1,2,4]triazolo[4,3-a]pyrazin-8-yl}oxy)pyridine-3-carboxylic acid dihydrochloride

  • MDL number MFCD22392237
  • Cas Number : 1421604-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112942
Availability details for
Quantity
  • Availability
  • Add to Cart