ENA372573531 Enamine

2-(4-nitro-1H-pyrazol-1-yl)ethanimidamide hydrochloride

  • MDL number MFCD22421790
  • Cas Number : 1423029-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113008
Availability details for
Quantity
  • Availability
  • Add to Cart