ENA372573533 Enamine

pyrimidin-2-ylmethanamine dihydrochloride

  • MDL number MFCD09997895
  • Cas Number : 1423031-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113029

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-113029
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart