ENA372573534 Enamine

5-(5-aminopyridin-3-yl)-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD22421792
  • Cas Number : 1423031-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113037

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113037
Availability details for
Quantity
  • Availability
  • Add to Cart