ENA372573536 Enamine

N-ethyl-1H-imidazol-2-amine hydrochloride

  • MDL number MFCD22421794
  • Cas Number : 1423034-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-113043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113043
Availability details for
Quantity
  • Availability
  • Add to Cart