ENA372573538 Enamine

4,4-difluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD22392240
  • Cas Number : 1421604-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113083
Availability details for
Quantity
  • Availability
  • Add to Cart