ENA372573540 Enamine

(1S)-1-(2-methyloxolan-2-yl)ethan-1-ol

  • MDL number MFCD22421797
  • Cas Number : 1423043-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113100
Availability details for
Quantity
  • Availability
  • Add to Cart