ENA372573546 Enamine

7-oxa-2-azaspiro[3.5]nonane hydrochloride

  • MDL number MFCD22421803
  • Cas Number : 1417633-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113203
Availability details for
Quantity
  • Availability
  • Add to Cart