ENA372573555 Enamine

9-[(thiophen-2-yl)methyl]-9-azabicyclo[3.3.1]nonan-3-amine dihydrochloride

  • MDL number MFCD22421812
  • Cas Number : 1423033-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113322
Availability details for
Quantity
  • Availability
  • Add to Cart