ENA372573558 Enamine

6-methylimidazo[2,1-b][1,3]thiazole-2-carboxylic acid hydrobromide

  • MDL number MFCD22421815
  • Cas Number : 1423034-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113336

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113336
Availability details for
Quantity
  • Availability
  • Add to Cart