ENA372573559 Enamine

2-(3,5-dimethylphenoxy)-1-(piperazin-1-yl)ethan-1-one hydrochloride

  • MDL number MFCD22421816
  • Cas Number : 1423033-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113339
Availability details for
Quantity
  • Availability
  • Add to Cart