ENA372573565 Enamine

1-(2-ethoxyethyl)cyclopentan-1-amine hydrochloride

  • MDL number MFCD22421824
  • Cas Number : 1423026-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113462

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113462
Availability details for
Quantity
  • Availability
  • Add to Cart