ENA372573571 Enamine

2-(2-bromo-5-chlorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD22421830
  • Cas Number : 1423025-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113513

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113513
Availability details for
Quantity
  • Availability
  • Add to Cart