ENA372573582 Enamine

2-(3,5-dichlorophenyl)-2-[(2,2,2-trifluoroethyl)amino]acetic acid hydrochloride

  • MDL number MFCD22421841
  • Cas Number : 1423026-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176...178
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113611

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113611
Availability details for
Quantity
  • Availability
  • Add to Cart