ENA372573588 Enamine

1-(4-fluorophenyl)-2-(pyrrolidin-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD22421847
  • Cas Number : 1423031-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162...164
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113693

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113693
Availability details for
Quantity
  • Availability
  • Add to Cart