ENA372573595 Enamine

3-[2-(4-methoxypyrrolidin-2-yl)-1H-imidazol-4-yl]pyridine trihydrochloride

  • MDL number MFCD22421853
  • Molecular Weight : 353.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-(4-methoxypyrrolidin-2-yl)-1H-imidazol-4-yl)pyridine trihydrochloride
Inchi Code 1S/C13H16N4O.3ClH/c1-18-10-5-11(15-7-10)13-16-8-12(17-13)9-3-2-4-14-6-9;;;/h2-4,6,8,10-11,15H,5,7H2,1H3,(H,16,17);3*1H
Inchi Key OJHSNXISIZYIEP-UHFFFAOYSA-N

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart