ENA372573615 Enamine

1-cyclopropyl-decahydro-1,6-naphthyridine

  • MDL number MFCD22421877
  • Cas Number : 1423031-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113998
Availability details for
Quantity
  • Availability
  • Add to Cart