ENA372573616 Enamine

2-(chloromethyl)-N-ethylpyridin-4-amine hydrochloride

  • MDL number MFCD22421878
  • Cas Number : 1423031-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114012
Availability details for
Quantity
  • Availability
  • Add to Cart