ENA372573622 Enamine

1-(5-fluoropyridin-2-yl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD22421885
  • Cas Number : 1421603-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114130
Availability details for
Quantity
  • Availability
  • Add to Cart