ENA372573625 Enamine

3-amino-2-[(3-methylphenyl)methyl]propanoic acid hydrochloride

  • MDL number MFCD22421890
  • Cas Number : 1423025-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114158
Availability details for
Quantity
  • Availability
  • Add to Cart