ENA372573626 Enamine

3-amino-2-[(2-methylphenyl)methyl]propanoic acid hydrochloride

  • MDL number MFCD22421891
  • Cas Number : 1423032-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114159
Availability details for
Quantity
  • Availability
  • Add to Cart