ENA372573627 Enamine

N-[3-(aminomethyl)phenyl]-2-(1H-pyrazol-1-yl)propanamide dihydrochloride

  • MDL number MFCD22421892
  • Cas Number : 1423031-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114167
Availability details for
Quantity
  • Availability
  • Add to Cart