ENA372573628 Enamine

N-(2-aminoethyl)pyridine-3-sulfonamide hydrochloride

  • MDL number MFCD22421893
  • Cas Number : 1423032-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114169

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114169
Availability details for
Quantity
  • Availability
  • Add to Cart