ENA372573630 Enamine

2-[1-(propan-2-yl)pyrrolidin-2-yl]acetic acid hydrochloride

  • MDL number MFCD22421895
  • Cas Number : 1423026-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114209
Availability details for
Quantity
  • Availability
  • Add to Cart