ENA372573632 Enamine

2-(5-chloro-1-benzothiophen-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD04150683
  • Cas Number : 23799-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-114232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114232
Availability details for
Quantity
  • Availability
  • Add to Cart