ENA372573634 Enamine

N-(2-aminoethyl)thiophene-2-sulfonamide hydrochloride

  • MDL number MFCD22421900
  • Cas Number : 1423032-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142...144
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114275

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114275
Availability details for
Quantity
  • Availability
  • Add to Cart