ENA372573650 Enamine

1-methyl-3-nitro-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD22421917
  • Cas Number : 1423033-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45531
Availability details for
Quantity
  • Availability
  • Add to Cart