ENA372573653 Enamine

methyl 1-(2-chloroacetamido)cyclohexane-1-carboxylate

  • MDL number MFCD22421920
  • Cas Number : 1423032-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46951
Availability details for
Quantity
  • Availability
  • Add to Cart