ENA372573656 Enamine

2-chloro-N-{[4-(2,6-dimethylmorpholin-4-yl)-3-fluorophenyl]methyl}acetamide

  • MDL number MFCD22392241
  • Cas Number : 1421603-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-48389

Safety Information

msds https://www.enaminestore.com/catalog/EN300-48389
Availability details for
Quantity
  • Availability
  • Add to Cart