ENA372573658 Enamine

1-(4-aminopiperidin-1-yl)-2-methylpropan-2-ol dihydrochloride

  • MDL number MFCD22392243
  • Cas Number : 1421602-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66349

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66349
Availability details for
Quantity
  • Availability
  • Add to Cart