ENA372573686 Enamine

2-[(oxan-4-yloxy)methyl]benzonitrile

  • MDL number MFCD14614952
  • Cas Number : 1249891-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113305
Availability details for
Quantity
  • Availability
  • Add to Cart