ENA372573692 Enamine

4,5,6,7-tetrahydro-1-benzothiophene-4-carboxylic acid

  • MDL number MFCD13192824
  • Cas Number : 339056-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113329
Availability details for
Quantity
  • Availability
  • Add to Cart