ENA372573695 Enamine

(2,2-dimethoxyethyl)(2-methylpropyl)amine

  • MDL number MFCD12148438
  • Cas Number : 1179210-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113333
Availability details for
Quantity
  • Availability
  • Add to Cart