ENA372573696 Enamine

(5S)-5-(hydroxymethyl)imidazolidine-2,4-dione

  • MDL number MFCD01076168
  • Cas Number : 40856-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113334
Availability details for
Quantity
  • Availability
  • Add to Cart