ENA372573701 Enamine

6'-methoxy-2',3'-dihydrospiro[imidazolidine-4,1'-indene]-2,5-dione

  • MDL number MFCD22340114
  • Cas Number : 66892-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113341

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113341
Availability details for
Quantity
  • Availability
  • Add to Cart