ENA372573705 Enamine

1-(2-bromoethyl)-3-(trifluoromethoxy)benzene

  • MDL number MFCD11846561
  • Cas Number : 958027-93-5
  • Molecular Weight : 269.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-bromoethyl)-3-(trifluoromethoxy)benzene
Inchi Code 1S/C9H8BrF3O/c10-5-4-7-2-1-3-8(6-7)14-9(11,12)13/h1-3,6H,4-5H2
Inchi Key RZHWLCIKYSVRGJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-113367

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-113367
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart