ENA372573708 Enamine

3-(2-amino-1,3-thiazol-4-yl)propanamide

  • MDL number MFCD22421819
  • Cas Number : 1105192-78-6
  • Molecular Weight : 171.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(2-aminothiazol-4-yl)propanamide
Inchi Code 1S/C6H9N3OS/c7-5(10)2-1-4-3-11-6(8)9-4/h3H,1-2H2,(H2,7,10)(H2,8,9)
Inchi Key TXUKUMHELBQYCI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-113371

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-113371
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart