ENA372573711 Enamine

N-(prop-2-yn-1-yl)-4-(trifluoromethyl)aniline

  • MDL number MFCD16664395
  • Cas Number : 233272-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113374
Availability details for
Quantity
  • Availability
  • Add to Cart