ENA372573716 Enamine

2-chloro-4-(dimethylamino)benzene-1-carboximidamide

  • MDL number MFCD17256877
  • Cas Number : 1340443-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112869
Availability details for
Quantity
  • Availability
  • Add to Cart