ENA372573717 Enamine

6-oxopiperidine-3-carboxamide

  • MDL number MFCD13178953
  • Cas Number : 189763-34-6
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113381
Availability details for
Quantity
  • Availability
  • Add to Cart