ENA372573725 Enamine

2-amino-N,3-dimethylbutanamide hydrochloride

  • MDL number MFCD13562338
  • Cas Number : 188890-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113403

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113403
Availability details for
Quantity
  • Availability
  • Add to Cart