ENA372573730 Enamine

4-[(4-bromo-1H-pyrazol-1-yl)methyl]-5-methylfuran-2-carboxylic acid

  • MDL number MFCD16729982
  • Cas Number : 1248793-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113410

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113410
Availability details for
Quantity
  • Availability
  • Add to Cart