ENA372573732 Enamine

imidazo[1,2-a]pyridine-7-carboxylic acid hydrochloride

  • MDL number MFCD09954929
  • Cas Number : 1423031-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 295-298
Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113423
Availability details for
Quantity
  • Availability
  • Add to Cart