ENA372573739 Enamine

N-(4-amino-3-methoxyphenyl)acetamide

  • MDL number MFCD03089241
  • Cas Number : 93973-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128...130
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113461

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113461
Availability details for
Quantity
  • Availability
  • Add to Cart