ENA372573741 Enamine

4-[(2-cyanophenyl)carbamoyl]butanoic acid

  • MDL number MFCD00090653
  • Cas Number : 197310-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112871

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112871
Availability details for
Quantity
  • Availability
  • Add to Cart