ENA372573742 Enamine

1-(2-methoxyethyl)-1H-pyrazole

  • MDL number MFCD18889154
  • Cas Number : 304693-68-3
  • Molecular Weight : 126.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-113464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113464
Availability details for
Quantity
  • Availability
  • Add to Cart