ENA372573749 Enamine

3-nitro-N-phenylaniline

  • MDL number MFCD00453770
  • Cas Number : 4531-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113472
Availability details for
Quantity
  • Availability
  • Add to Cart