ENA372573750 Enamine

3-cyano-4-ethylbenzoic acid

  • MDL number MFCD20658809
  • Cas Number : 684249-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113473
Availability details for
Quantity
  • Availability
  • Add to Cart